2-(2-Formyl-6-methoxyphenoxy)acetonitrile 95%
Dodavatel: Apollo Scientific
OR33272-1G
OR33272-25G
OR33272-5G
APOSOR33272-1GEA
5130
CZK
APOSOR33272-1G
APOSOR33272-25G
APOSOR33272-5G
2-(2-Formyl-6-methoxyphenoxy)acetonitrile 95%
2-(2-Formyl-6-methoxyphenoxy)acetonitrile
Vzorec:
C₁₀H₉NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD02082429 CAS číslo: 127500-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...