3-Hydroxypyrido[1,2-a]indole-10-carbonitrile 95%
Dodavatel: Apollo Scientific
OR33263-1G
OR33263-5G
APOSOR33263-1GEA
6950
CZK
APOSOR33263-1G
APOSOR33263-5G
3-Hydroxypyrido[1,2-a]indole-10-carbonitrile 95%
3-Hydroxypyrido[1,2-a]indole-10-carbonitrile
Vzorec:
C₁₃H₈N₂O MW: 208.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01568483 CAS číslo: 339106-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...