2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile 95%
Dodavatel: Apollo Scientific
OR33256-1G
OR33256-5G
APOSOR33256-1GEA
3480
CZK
APOSOR33256-1G
APOSOR33256-5G
2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile 95%
2-[4-(2-Pyrimidinyloxy)phenyl]acetonitrile
Vzorec:
C₁₂H₉N₃O MW: 211,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01443818 CAS číslo: 339105-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...