Tisk…

4-[(2-Chloro-1,3-thiazol-5-yl)methyl]morpholine 95%

Dodavatel: Apollo Scientific

OR33253-1G OR33253-5G
APOSOR33253-1GEA 7670 CZK
APOSOR33253-1G APOSOR33253-5G
4-[(2-Chloro-1,3-thiazol-5-yl)methyl]morpholine 95%
4-[(2-Chloro-1,3-thiazol-5-yl)methyl]morpholine
Vzorec: C₈H₁₁ClN₂OS
Storage Temperature: Ambient
MDL Number: MFCD01443905
CAS číslo: 339105-10-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR