Tisk…

2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-phenylpropanoic acid 95%

Dodavatel: Apollo Scientific

OR33238-1G OR33238-25G OR33238-5G
APOSOR33238-1GEA 5130 CZK
APOSOR33238-1G APOSOR33238-25G APOSOR33238-5G
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-phenylpropanoic acid 95%
2-(1-Oxo-1,3-dihydro-2H-isoindol-2-yl)-3-phenylpropanoic acid
Vzorec: C₁₇H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00664644
CAS číslo: 96017-10-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR