Tisk…

Ethyl-3-(2-methyl-1H-imidazol-1-yl)propanoate 95%

Dodavatel: Apollo Scientific

OR33236-1G
APOSOR33236-1GEA 7670 CZK
APOSOR33236-1G
Ethyl-3-(2-methyl-1H-imidazol-1-yl)propanoate 95%
Ethyl-3-(2-methyl-1H-imidazol-1-yl)propanoate
Vzorec: C₉H₁₄N₂O₂
MW: 182,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02602292
CAS číslo: 18999-48-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR