Tisk…

(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(2-Chloropyridine-3-carbonyl)morpholine, (2-Chloropyridin-3-yl)(morpholino)methanone

OR33225-1G OR33225-5G
APOSOR33225-1GEA 5790 CZK
APOSOR33225-1G APOSOR33225-5G
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone 95%
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone
Vzorec: C₁₀H₁₁ClN₂O₂
MW: 226,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00454210
CAS číslo: 53062-98-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR