2,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide 95%
Dodavatel: Apollo Scientific
Synonyms:
2,4-Dichloro-N-(2-hydroxyethyl)benzamide
OR33223-1G
OR33223-5G
APOSOR33223-1GEA
5130
CZK
APOSOR33223-1G
APOSOR33223-5G
2,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide 95%
2,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide
Vzorec:
C₉H₉Cl₂NO₂ MW: 234,08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00141691 CAS číslo: 20656-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...