Tisk…

4-(1,3-Benzodioxol-5-yl)benzenecarboxylic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(2H-1,3-Benzodioxol-5-yl)benzoic acid

OR33199-1G OR33199-5G
APOSOR33199-1GEA 15310 CZK
APOSOR33199-1G APOSOR33199-5G
4-(1,3-Benzodioxol-5-yl)benzenecarboxylic acid 95%
4-(1,3-Benzodioxol-5-yl)benzenecarboxylic acid
Vzorec: C₁₄H₁₀O₄
MW: 242,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426480
CAS číslo: 193151-97-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR