Tisk…

Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: Methyl 5-bromo-2-methoxy-3-methylbenzoate

OR33194-1G OR33194-5G
APOSOR33194-1GEA 7670 CZK
APOSOR33194-1G APOSOR33194-5G
Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate 95%
Methyl-5-bromo-2-methoxy-3-methylbenzenecarboxylate
Vzorec: C₁₀H₁₁BrO₃
MW: 259,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659589
CAS číslo: 722497-32-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR