Tisk…

4-(4-Methoxyphenyl)-4-oxobutyronitrile 95%

Dodavatel: Apollo Scientific

OR33190-1G
APOSOR33190-1GEA 9660 CZK
APOSOR33190-1G
4-(4-Methoxyphenyl)-4-oxobutyronitrile 95%
4-(4-Methoxyphenyl)-4-oxobutyronitrile
Vzorec: C₁₁H₁₁NO₂
MW: 189,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00158889
CAS číslo: 55234-56-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR