Tisk…

Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride 95%

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-amino-2-(4-bromophenyl)acetate hydrochloride

OR33179-1G OR33179-5G
APOSOR33179-1GEA 2240 CZK
APOSOR33179-1G APOSOR33179-5G
Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride 95%
Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride
Vzorec: C₉H₁₁BrClNO₂
MW: 280,56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663802
CAS číslo: 42718-20-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR