Tisk…

1-(3-Amino-4-piperidinophenyl)-1-ethanone 95%

Dodavatel: Apollo Scientific

OR33178-1G OR33178-5G
APOSOR33178-1GEA 8700 CZK
APOSOR33178-1G APOSOR33178-5G
1-(3-Amino-4-piperidinophenyl)-1-ethanone 95%
1-(3-Amino-4-piperidinophenyl)-1-ethanone
Vzorec: C₁₃H₁₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD00116065
CAS číslo: 30877-81-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR