Tisk…

1-(3-Amino-4-morpholinophenyl)-1-ethanone 95%

Dodavatel: Apollo Scientific

OR33177-1G OR33177-5G
APOSOR33177-1GEA 15310 CZK
APOSOR33177-1G APOSOR33177-5G
1-(3-Amino-4-morpholinophenyl)-1-ethanone 95%
1-(3-Amino-4-morpholinophenyl)-1-ethanone
Vzorec: C₁₂H₁₆N₂O₂
MW: 220,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00116066
CAS číslo: 217489-78-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR