6-(1-Pyrrolidinyl)nicotinic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
6-(Pyrrolidin-1-yl)pyridine-3-carboxylic acid, 6-Pyrrolidin-1-yl-nicotinic acid
OR33166-1G
OR33166-5G
APOSOR33166-1GEA
9560
CZK
APOSOR33166-1G
APOSOR33166-5G
6-(1-Pyrrolidinyl)nicotinic acid 95%
6-(1-Pyrrolidinyl)nicotinic acid
Vzorec:
C₁₀H₁₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03791234 CAS číslo: 210963-95-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...