Tisk…

N-(4-Methylbenzyl)urea 95%

Dodavatel: Apollo Scientific

OR33121-1G OR33121-25G OR33121-5G
APOSOR33121-1GEA 5130 CZK
APOSOR33121-1G APOSOR33121-25G APOSOR33121-5G
N-(4-Methylbenzyl)urea 95%
N-(4-Methylbenzyl)urea
Vzorec: C₉H₁₂N₂O
MW: 164.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01314625
CAS číslo: 54582-34-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR