Tisk…

3-Cyanophenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Cyanophenyl)metaboric acid, Dihydroxy(3-cyanophenyl)borane, 3-Cyanobenzeneboronic acid

OR3312-25G OR3312-5G
APOSOR3312-25GEA 992 CZK
APOSOR3312-25G APOSOR3312-5G
3-Cyanophenylboronic acid 98%
3-Cyanophenylboronic acid
Vzorec: C₇H₆BNO₂
MW: 146,94 g/mol
Bod tání: 298 °C
Storage Temperature: Ambient
MDL Number: MFCD01318967
CAS číslo: 150255-96-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR