Tisk…

4-Chloro-N-(5-chloro-2-pyridinyl)butanamide 95%

Dodavatel: Apollo Scientific

OR33101-1G OR33101-5G OR33101-25G
APOSOR33101-1GEA 5130 CZK
APOSOR33101-1G APOSOR33101-5G APOSOR33101-25G
4-Chloro-N-(5-chloro-2-pyridinyl)butanamide 95%
4-Chloro-N-(5-chloro-2-pyridinyl)butanamide
Vzorec: C₉H₁₀Cl₂N₂O
MW: 233,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664609
CAS číslo: 339014-78-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR