Tisk…

(E)-3-(Dimethylamino)-1-(2-naphthyl)-2-propen-1-one 95%

Dodavatel: Apollo Scientific

OR33098-1G OR33098-5G
APOSOR33098-1GEA 5130 CZK
APOSOR33098-1G APOSOR33098-5G
(E)-3-(Dimethylamino)-1-(2-naphthyl)-2-propen-1-one 95%
(E)-3-(Dimethylamino)-1-(2-naphthyl)-2-propen-1-one
Vzorec: C₁₅H₁₅NO
MW: 225,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00179142
CAS číslo: 138716-23-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR