Tisk…
2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride 95%
Dodavatel: Apollo Scientific
OR33096-1G OR33096-5G
APOSOR33096-1GEA 9560 CZK
APOSOR33096-1G APOSOR33096-5G
2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride 95%
2-Amino-1-(2-naphthyl)-1-ethanone hydrochloride
Vzorec: C₁₂H₁₂ClNO
MW: 221.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056659
CAS číslo: 38061-36-8