Tisk…

6-Methyl-3-(4-methylphenyl)-1,2,4-triazin-5-ol 95%

Dodavatel: Apollo Scientific

OR33095-1G OR33095-5G
APOSOR33095-1GEA 9560 CZK
APOSOR33095-1G APOSOR33095-5G
6-Methyl-3-(4-methylphenyl)-1,2,4-triazin-5-ol 95%
6-Methyl-3-(4-methylphenyl)-1,2,4-triazin-5-ol
Vzorec: C₁₁H₁₁N₃O
Storage Temperature: Ambient
MDL Number: MFCD04117816
CAS číslo: 109306-98-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR