Tisk…

6-Indoleboronic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Indole-6-boronic acid

OR3309-1G OR3309-5G OR3309-25G
APOSOR3309-1GEA 1240 CZK
APOSOR3309-1G APOSOR3309-5G APOSOR3309-25G
6-Indoleboronic acid 95%
6-Indoleboronic acid
Vzorec: C₈H₈BNO₂
MW: 160,97 g/mol
Bod tání: 177…181 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095176
CAS číslo: 147621-18-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR