Ethyl-4-hydroxy-7,8-dimethylquinoline-3-carboxylate 95%
Dodavatel: Apollo Scientific
OR33089-1G
OR33089-5G
APOSOR33089-1GEA
5130
CZK
APOSOR33089-1G
APOSOR33089-5G
Ethyl-4-hydroxy-7,8-dimethylquinoline-3-carboxylate 95%
Ethyl-4-hydroxy-7,8-dimethylquinoline-3-carboxylate
Vzorec:
C₁₄H₁₅NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00665075 CAS číslo: 53164-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...