Tisk…

Methyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate 95%

Dodavatel: Apollo Scientific

OR33051-1G OR33051-5G
APOSOR33051-1GEA 5130 CZK
APOSOR33051-1G APOSOR33051-5G
Methyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate 95%
Methyl-2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate
Vzorec: C₇H₁₁N₃O₂
MW: 169,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659569
CAS číslo: 885949-78-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR