(1-Benzyl-3-piperidinyl)methanol 95%
Dodavatel: Apollo Scientific
OR33044-1G
OR33044-5G
OR33044-25G
APOSOR33044-1GEA
3480
CZK
APOSOR33044-1G
APOSOR33044-5G
APOSOR33044-25G
(1-Benzyl-3-piperidinyl)methanol 95%
(1-Benzyl-3-piperidinyl)methanol
Vzorec:
C₁₃H₁₉NO Storage Temperature: Ambient |
CAS číslo:
85387-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...