Tisk…
2,6-Dibromopyridine N-Oxide 97%
Dodavatel: Apollo Scientific
Synonyms: 2,6-Dibromopyridine 1-oxide, 2,6-Dibromopyridin-1-ium-1-olate
OR33007-1G OR33007-5G
APOSOR33007-1GEA 2030 CZK
APOSOR33007-1G APOSOR33007-5G
2,6-Dibromopyridine N-Oxide 97%
2,6-Dibromopyridine N-Oxide
Vzorec: C₅H₃Br₂NO
MW: 252.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234019
CAS číslo: 25373-69-7