Tisk…

[5-Phenyl-3-isoxazolyl]methanol 95%

Dodavatel: Apollo Scientific

Synonyms: (5-Phenylisoxazol-3-yl)methanol, 3-Hydroxymethyl-5-phenylisoxazole

OR33005-1G OR33005-5G
APOSOR33005-1GEA 5130 CZK
APOSOR33005-1G APOSOR33005-5G
[5-Phenyl-3-isoxazolyl]methanol 95%
[5-Phenyl-3-isoxazolyl]methanol
Vzorec: C₁₀H₉NO₂
MW: 175,1840 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01444104
CAS číslo: 1619-37-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR