Ethyl-8-chloro[1,3]dioxolo[4,5-g]quinoline-7-carboxylate 95%
Dodavatel: Apollo Scientific
OR32982-1G
APOSOR32982-1GEA
6390
CZK
APOSOR32982-1G
Ethyl-8-chloro[1,3]dioxolo[4,5-g]quinoline-7-carboxylate 95%
Ethyl-8-chloro[1,3]dioxolo[4,5-g]quinoline-7-carboxylate
Vzorec:
C₁₃H₁₀ClNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00173405 CAS číslo: 26893-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...