Tisk…

3-[2-(1H-Imidazol-1-yl)phenyl]-1H-1,2,4-triazole 95%

Dodavatel: Apollo Scientific

OR32974-1G OR32974-5G
APOSOR32974-1GEA 9560 CZK
APOSOR32974-1G APOSOR32974-5G
3-[2-(1H-Imidazol-1-yl)phenyl]-1H-1,2,4-triazole 95%
3-[2-(1H-Imidazol-1-yl)phenyl]-1H-1,2,4-triazole
Vzorec: C₁₁H₉N₅
Storage Temperature: Ambient
MDL Number: MFCD00244275
CAS číslo: 338793-57-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR