Tisk…

2-(Allyloxy)-3-methoxybenzenecarbaldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Methoxy-2-(prop-2-en-1-yloxy)benzaldehyde

OR32970-1G
APOSOR32970-1GEA 5860 CZK
APOSOR32970-1G
2-(Allyloxy)-3-methoxybenzenecarbaldehyde 95%
2-(Allyloxy)-3-methoxybenzenecarbaldehyde
Vzorec: C₁₁H₁₂O₃
MW: 192,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03001271
CAS číslo: 23343-06-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR