Tisk…

3-(2,5-Dimethyl-1H-pyrrol-1-yl)propanoic acid 95%

Dodavatel: Apollo Scientific

OR32947-1G OR32947-5G
APOSOR32947-1GEA 7670 CZK
APOSOR32947-1G APOSOR32947-5G
3-(2,5-Dimethyl-1H-pyrrol-1-yl)propanoic acid 95%
3-(2,5-Dimethyl-1H-pyrrol-1-yl)propanoic acid
Vzorec: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD02223275
CAS číslo: 134796-54-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR