Tisk…

Ethyl-1-(4-chlorobenzoyl)-4-piperidinecarboxylate 95%

Dodavatel: Apollo Scientific

OR32913-1G OR32913-5G OR32913-25G
APOSOR32913-1GEA 5130 CZK
APOSOR32913-1G APOSOR32913-5G APOSOR32913-25G
Ethyl-1-(4-chlorobenzoyl)-4-piperidinecarboxylate 95%
Ethyl-1-(4-chlorobenzoyl)-4-piperidinecarboxylate
Vzorec: C₁₅H₁₈ClNO₃
MW: 295,77 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01213111
CAS číslo: 349397-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR