Tisk…

1-Amino-2-methylnaphthalene 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Methylnaphthalen-1-amine, 2-Methyl-1-naphthylamine

OR32890-1G OR32890-5G
APOSOR32890-1GEA 827 CZK
APOSOR32890-1G APOSOR32890-5G
1-Amino-2-methylnaphthalene 95%
1-Amino-2-methylnaphthalene
Vzorec: C₁₁H₁₁N
MW: 157,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059135
CAS číslo: 2246-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR