Tisk…

2-(Allyloxy)-5-chlorobenzaldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: 5-Chloro-2-(prop-2-en-1-yloxy)benzaldehyde

OR32851-1G OR32851-25G OR32851-5G
APOSOR32851-1GEA 5130 CZK
APOSOR32851-1G APOSOR32851-25G APOSOR32851-5G
2-(Allyloxy)-5-chlorobenzaldehyde 95%
2-(Allyloxy)-5-chlorobenzaldehyde
Vzorec: C₁₀H₉ClO₂
MW: 196,63 g/mol
Storage Temperature: Ambient
CAS číslo: 152842-93-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR