1-(2,6-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
1-[(2,6-Dichlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid
OR32829-1G
OR32829-5G
OR32829-25G
APOSOR32829-1GEA
3480
CZK
APOSOR32829-1G
APOSOR32829-5G
APOSOR32829-25G
1-(2,6-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid 95%
1-(2,6-Dichlorobenzyl)-2-oxo-1,2-dihydro-3-pyridine carboxylic acid
Vzorec:
C₁₃H₉Cl₂NO₃ MW: 298,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00139966 CAS číslo: 338754-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...