2-Iodophenyl acetate 95%
Dodavatel: Apollo Scientific
OR32806-1G
OR32806-25G
OR32806-5G
APOSOR32806-1GEA
5130
CZK
APOSOR32806-1G
APOSOR32806-25G
APOSOR32806-5G
2-Iodophenyl acetate 95%
2-Iodophenyl acetate
Vzorec:
C₈H₇IO₂ MW: 262,0445 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07780625 CAS číslo: 32865-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...