6-(4-Benzylpiperazino)nicotinaldehyde 95%
Dodavatel: Apollo Scientific
Synonyms:
6-(4-Benzylpiperazin-1-yl)pyridine-3-carbaldehyde
OR32797-1G
OR32797-5G
APOSOR32797-1GEA
15310
CZK
APOSOR32797-1G
APOSOR32797-5G
6-(4-Benzylpiperazino)nicotinaldehyde 95%
6-(4-Benzylpiperazino)nicotinaldehyde
Vzorec:
C₁₇H₁₉N₃O MW: 281,36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973728 CAS číslo: 886360-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...