Tisk…

5-Methyl-2-furaldehyde oxime 95%

Dodavatel: Apollo Scientific

OR32786-1G OR32786-25G OR32786-5G
APOSOR32786-1GEA 5130 CZK
APOSOR32786-1G APOSOR32786-25G APOSOR32786-5G
5-Methyl-2-furaldehyde oxime 95%
5-Methyl-2-furaldehyde oxime
Vzorec: C₆H₇NO₂
MW: 125,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00482440
CAS číslo: 32750-36-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR