Tisk…

3-(tert-Butoxy)-6-chloropyridazine 95%

Dodavatel: Apollo Scientific

OR32780-5G
APOSOR32780-5GEA 3610 CZK
APOSOR32780-5G
3-(tert-Butoxy)-6-chloropyridazine 95%
3-(tert-Butoxy)-6-chloropyridazine
Vzorec: C₈H₁₁ClN₂O
MW: 186.6400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425769
CAS číslo: 17321-24-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR