2-(4-Piperidinyl)-1,3-benzothiazole 95%
Dodavatel: Apollo Scientific
OR32774-1G
APOSOR32774-1GEA
5130
CZK
APOSOR32774-1G
2-(4-Piperidinyl)-1,3-benzothiazole 95%
2-(4-Piperidinyl)-1,3-benzothiazole
Vzorec:
C₁₂H₁₄N₂S MW: 218,32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01815501 CAS číslo: 51784-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...