Tisk…

2-Chloro-3-oxo-N-phenylbutanamide 95%

Dodavatel: Apollo Scientific

OR32769-1G OR32769-5G
APOSOR32769-1GEA 3740 CZK
APOSOR32769-1G APOSOR32769-5G
2-Chloro-3-oxo-N-phenylbutanamide 95%
2-Chloro-3-oxo-N-phenylbutanamide
Vzorec: C₁₀H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD01838235
CAS číslo: 31844-92-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR