Methyl[(4-methylphenyl)methylene]ammoniumolate 95%
Dodavatel: Apollo Scientific
OR32753-1G
OR32753-5G
APOSOR32753-1GEA
7670
CZK
APOSOR32753-1G
APOSOR32753-5G
Methyl[(4-methylphenyl)methylene]ammoniumolate 95%
Methyl[(4-methylphenyl)methylene]ammoniumolate
Vzorec:
C₉H₁₁NO Storage Temperature: Ambient |
MDL Number:
MFCD00231569 CAS číslo: 16089-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...