6-Methoxy-2H-chromene-3-carbonitrile 95%
Dodavatel: Apollo Scientific
OR32729-1G
OR32729-5G
APOSOR32729-1GEA
4630
CZK
APOSOR32729-1G
APOSOR32729-5G
6-Methoxy-2H-chromene-3-carbonitrile 95%
6-Methoxy-2H-chromene-3-carbonitrile
Vzorec:
C₁₁H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00202032 CAS číslo: 57543-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...