Tisk…

8-Methoxy-2H-chromene-3-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR32723-1G OR32723-5G
APOSOR32723-1GEA 6950 CZK
APOSOR32723-1G APOSOR32723-5G
8-Methoxy-2H-chromene-3-carboxylic acid 95%
8-Methoxy-2H-chromene-3-carboxylic acid
Vzorec: C₁₁H₁₀O₄
Storage Temperature: Ambient
CAS číslo: 57543-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR