3-Bromo-2-formylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Bromopyridine-2-carboxaldehyde
OR3272-1G
OR3272-500MG
APOSOR3272-1GEA
1740
CZK
APOSOR3272-1G
APOSOR3272-500MG
3-Bromo-2-formylpyridine
3-Bromo-2-formylpyridine
Vzorec:
C₆H₄BrNO MW: 186,01 g/mol Bod tání: 62…63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07781220 CAS číslo: 405174-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...