(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol 97%
Dodavatel: Apollo Scientific
OR32719-1G
OR32719-5G
OR32719-25G
APOSOR32719-1GEA
3770
CZK
APOSOR32719-1G
APOSOR32719-5G
APOSOR32719-25G
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol 97%
(1-Phenyl-1H-1,2,3-triazol-4-yl)methanol
Vzorec:
C₉H₉N₃O MW: 175,1900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00100214 CAS číslo: 103755-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...