Methyl-2-(cyanomethoxy)benzenecarboxylate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 2-(cyanomethoxy)benzoate
OR32715-1G
OR32715-5G
APOSOR32715-1GEA
3480
CZK
APOSOR32715-1G
APOSOR32715-5G
Methyl-2-(cyanomethoxy)benzenecarboxylate 95%
Methyl-2-(cyanomethoxy)benzenecarboxylate
Vzorec:
C₁₀H₉NO₃ MW: 191,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01924694 CAS číslo: 1641-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...