Tisk…

3-(3-Phenyl-2-propenyl)-2,4-pentanedione 95%

Dodavatel: Apollo Scientific

OR32675-1G OR32675-5G
APOSOR32675-1GEA 7670 CZK
APOSOR32675-1G APOSOR32675-5G
3-(3-Phenyl-2-propenyl)-2,4-pentanedione 95%
3-(3-Phenyl-2-propenyl)-2,4-pentanedione
Vzorec: C₁₄H₁₆O₂
MW: 216,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425758
CAS číslo: 29638-71-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR