Tisk…

Ethyl 2-cyclohexylidenacetate 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-cyclohexylideneacetate

OR32656-1G OR32656-5G
APOSOR32656-1GEA 870 CZK
APOSOR32656-1G APOSOR32656-5G
Ethyl 2-cyclohexylidenacetate 95%
Ethyl-2-cyclohexylidenacetate
Vzorec: C₁₀H₁₆O₂
MW: 168.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00963857
CAS číslo: 1552-92-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR