Tisk…

6-(Methoxymethyl)-2-methyl-4-pyrimidinol 95%

Dodavatel: Apollo Scientific

OR32640-1G OR32640-25G OR32640-5G
APOSOR32640-1GEA 5130 CZK
APOSOR32640-1G APOSOR32640-25G APOSOR32640-5G
6-(Methoxymethyl)-2-methyl-4-pyrimidinol 95%
6-(Methoxymethyl)-2-methyl-4-pyrimidinol
Vzorec: C₇H₁₀N₂O₂
MW: 154,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00087858
CAS číslo: 38249-50-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR